Sivelestat neutrophil elastase inhibitor
À propos de cet article
Potent, selective, substrate competitive neutrophil elastase inhibitor (IC₅₀ values are 19 to 49 nM and 5.6 μM for leukocyte and pancreas elastase respectively). Suppresses neutrophil-induced capillary permeability and leukocyte kinetics. Active in vivo.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:127373-66-4
- Molecular Weight:434.5 g/mol
2 Options Available Below
Product Details & Documents
Potent, selective, substrate competitive neutrophil elastase inhibitor (IC₅₀ values are 19 to 49 nM and 5.6 μM for leukocyte and pancreas elastase respectively). Suppresses neutrophil-induced capillary permeability and leukocyte kinetics. Active in vivo.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:127373-66-4
- Molecular Weight:434.5 g/mol
Specifications for Product Family
Résultats des tests de spécification
- Constituents::100% Sivelestat



