Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Procainamide hydrochloride ≥99% Sodium channel blocker

Procainamide hydrochloride ≥99% Sodium channel blocker

New Product
Supplier:   Abcam
Procainamide hydrochloride ≥99% Sodium channel blocker
Supplier:  Abcam
New Product
Certificates
Procainamide hydrochloride ≥99% Sodium channel blocker

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sodium channel blocker. Specific DNA methyltransferase 1 inhibitor (Ki =7.2 μM). Antiarrhythmic in vivo. Limited ability to cross the blood-brain barrier.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  4-Amino-N-[2'-(diethylamino)ethyl]benzamide.HCl
  • CAS Number:614-39-1
  • Molecular Weight:271.79 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Procainamide hydrochloride
Product Family Options

Product Information

  • Taille
    Availability
    Price
  • 10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $113.00
    $113.00
    Each (10g)
       $113.00

    Supplier Number: AB120955-10G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • 25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $195.00
    $195.00
    Each (25g)
       $195.00
    Minimum order quantity is 1

    Supplier Number: AB120955-25G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.