Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Diazoxide ≥99% KIR6.x (ATP-sensitive K⁺ channel) activator | Avantor

Diazoxide ≥99% KIR6.x (ATP-sensitive K⁺ channel) activator

100 mg
Catalog # ABCA_AB120266-100M
New Product
Supplier:   Abcam
Fournisseur #:  AB120266-100MG
Diazoxide ≥99% KIR6.x (ATP-sensitive K⁺ channel) activator
Catalog # ABCA_AB120266-100M
Supplier:  Abcam
New Product
CAS Number:  364-98-7
Diazoxide ≥99% KIR6.x (ATP-sensitive K⁺ channel) activator

100 mg

CAS Number:  364-98-7
Price
$133.00
Log in pour la disponibilité
Each (100mg)
La quantité minimale de commande est de 1
Product Details & Documents

Selective KIR6.x (KATP) activator; antihypertensive. Achieve your results faster with highly validated, pure and trusted compounds.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonymes:  3-Methyl-7-chloro-1,2,4-benzothiadiazine 1,1-dioxide, 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-Dioxide
  • CAS Number:364-98-7
  • Molecular Weight:230.67 g/mol
Spécifications

Résultats des tests de spécification

  • Constituents::
    100% Diazoxide

Spécifications

  • Taille:
    100 mg

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.