pH Calibration Buffer Solution
pH 12.45 calibration buffer solution (230 ml)
Catalog # 470359-708
New Product
Supplier: MILWAUKEE INSTRUMENTS, INC.
Supplier #: MA9112
pH Calibration Buffer Solution
Catalog # 470359-708
Supplier: MILWAUKEE INSTRUMENTS, INC.
New Product
CAS Number:
Jump to:
Product Details & Documents
Premium quality, high accuracy pH buffer for calibrating pH meters.
Milwaukee’s buffer solution is laboratory grade and certified to meet the standard to an accuracy of ±0.01 pH removing the inaccuracies involved with mixing buffers
- Fundamental for the correct use of electrodes and for obtaining the most accurate and reproducible readings
- Sealed against light with airtight, tamper proof cap
- Ideal for fast accurate calibration
Milwaukee calibration buffers have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Milwaukee's line of calibration buffers have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle.
Specifications
Specifications
- Désignation:pH 12.45 calibration buffer solution (230 ml)



