Cell Strainers, Sterile
New Product
Supplier: CELLTREAT Scientific Products
Jump to:
À propos de cet article
Cell strainers for isolating single cells and removing debris or aggregate from cell suspensions.
- Can be used for many applications including isolating cells and removing clumps or debris
Hydrophilic treatment increases flow and reduces the effects of surface tension
Designed to universally fit most brands of 50 ml conical tubes
Extended lip on housing enables aseptic handling with forceps
7 Options Available Below
Product Details & Documents
Cell strainers for isolating single cells and removing debris or aggregate from cell suspensions.
- Can be used for many applications including isolating cells and removing clumps or debris
Hydrophilic treatment increases flow and reduces the effects of surface tension
Designed to universally fit most brands of 50 ml conical tubes
Extended lip on housing enables aseptic handling with forceps
Product Family Options
Informations sur le produit
- DésignationSterilityColorMatièrePackagingAvailabilityPrice
- Pestle, cell strainerSterileGreenPolypropyleneIndividually wrappedLog in pour la disponibilitéCase of 50$114.54Supplier Number: 229480Spécifications:New ProductNew Product
- Cell strainer, 40 μmSterileBluePolypropyleneIndividually wrappedLog in pour la disponibilitéCase of 50$264.41Supplier Number: 229481Spécifications:New ProductNew Product
- Cell strainer, 40 μmSterileBluePolypropyleneBulk packedLog in pour la disponibilitéCase of 50$254.94Supplier Number: 229482Spécifications:New ProductNew Product
- Cell strainer, 70 μmSterileWhitePolypropyleneIndividually wrappedLog in pour la disponibilitéCase of 50$264.41Supplier Number: 229483Spécifications:New ProductNew Product
- Cell strainer, 70 μmSterileWhitePolypropyleneBulk packedLog in pour la disponibilitéCase of 50$254.94Supplier Number: 229484Spécifications:New ProductNew Product
- Cell strainer, 100 μmSterileYellowPolypropyleneIndividually wrappedLog in pour la disponibilitéCase of 50$264.41Supplier Number: 229485Spécifications:New ProductNew Product
- Cell strainer, 100 μmSterileYellowPolypropyleneBulk packedLog in pour la disponibilitéCase of 50$254.94Supplier Number: 229486Spécifications:New ProductNew Product