Cell Strainers, Sterile
New Product
Cell Strainers, Sterile
New Product
Certificates
Cell Strainers, Sterile
Cell Strainers, Sterile
7 Options Available Below
Product Details & Documents
Cell strainers for isolating single cells and removing debris or aggregate from cell suspensions.
  • Can be used for many applications including isolating cells and removing clumps or debris
    Hydrophilic treatment increases flow and reduces the effects of surface tension
    Designed to universally fit most brands of 50 ml conical tubes
    Extended lip on housing enables aseptic handling with forceps
Product Family Options
Informations sur le produit
  • Désignation
    Sterility
    Color
    Matière
    Packaging
    Availability
    Price
  • Pestle, cell strainer
    Sterile
    Green
    Polypropylene
    Individually wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $114.54
    $114.54
    Case of 50
       $114.54
    Supplier Number: 229480
    Spécifications:
    New Product
    New Product
  • Cell strainer, 40 μm
    Sterile
    Blue
    Polypropylene
    Individually wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $264.41
    $264.41
    Case of 50
       $264.41
    Supplier Number: 229481
    Spécifications:
    New Product
    New Product
  • Cell strainer, 40 μm
    Sterile
    Blue
    Polypropylene
    Bulk packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $254.94
    $254.94
    Case of 50
       $254.94
    Supplier Number: 229482
    Spécifications:
    New Product
    New Product
  • Cell strainer, 70 μm
    Sterile
    White
    Polypropylene
    Individually wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $264.41
    $264.41
    Case of 50
       $264.41
    Supplier Number: 229483
    Spécifications:
    New Product
    New Product
  • Cell strainer, 70 μm
    Sterile
    White
    Polypropylene
    Bulk packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $254.94
    $254.94
    Case of 50
       $254.94
    Supplier Number: 229484
    Spécifications:
    New Product
    New Product
  • Cell strainer, 100 μm
    Sterile
    Yellow
    Polypropylene
    Individually wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $264.41
    $264.41
    Case of 50
       $264.41
    Supplier Number: 229485
    Spécifications:
    New Product
    New Product
  • Cell strainer, 100 μm
    Sterile
    Yellow
    Polypropylene
    Bulk packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $254.94
    $254.94
    Case of 50
       $254.94
    Supplier Number: 229486
    Spécifications:
    New Product
    New Product