Plasteur® Pasteur Pipets
Plasteur® Pasteur Pipets
New Product
Plasteur® Pasteur Pipets
New Product
Certificates
Plasteur® Pasteur Pipets
Plasteur® Pasteur Pipets
13 Options Available Below
Product Details & Documents
Plasteur pipets for transferring liquids.
  • CELLTREAT® exclusive product
    Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
    Flexible plastic helps prevent glove or skin punctures
    Available packaged sterile individually wrapped or bulk packed
    Available packaged non-sterile in re-sealable bags or tab lock boxes
    Eliminate sharps disposal costs
Product Family Options
Polystyrene Plasteur® Pipets
Product Information
  • Désignation
    Longueur
    Availability
    Price
  • Individually wrapped, sterile
    5.75"
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $159.58
    $159.58
    Case of 200
       $159.58
    Supplier Number: 229270
    Specifications:
    New Product
    New Product
  • Bulk packed, sterile
    5.75"
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $339.37
    $339.37
    Case of 500
       $339.37
    Supplier Number: 229275
    Specifications:
    New Product
    New Product
  • Individually wrapped, sterile
    9"
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $195.92
    $195.92
    Case of 200
       $195.92
    Supplier Number: 229280
    Specifications:
    New Product
    New Product
  • Bulk packed, sterile
    9"
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $374.46
    $374.46
    Case of 500
       $374.46
    Supplier Number: 229285
    Specifications:
    New Product
    New Product
Polypropylene Plasteur® Pipets
Product Information
  • Désignation
    Longueur
    Availability
    Price
  • Individually wrapped, sterile
    5.75"
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $214.88
    $214.88
    Case of 200
       $214.88
    Supplier Number: 229271
    Specifications:
    New Product
    New Product
  • Bulk Packed, tab lock box, non-sterile
    5.75"
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $270.18
    $270.18
    Case of 1,000
       $270.18
    Supplier Number: 229274
    Specifications:
    New Product
    New Product
  • Bulk packed, sterile
    5.75"
    Log in for availability
    Log in for availability
    Each
    Each - $255.96
    $255.96
    Each
       $255.96
    Supplier Number: 229276
    Specifications:
    New Product
    New Product
  • Bulk packed, non-sterile
    5.75"
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $194.34
    $194.34
    Case of 500
       $194.34
    Supplier Number: 229277
    Specifications:
    New Product
    New Product
  • Individually wrapped, sterile
    9"
    Log in for availability
    Log in for availability
    Case of 200
    Case of 200 - $222.78
    $222.78
    Case of 200
       $222.78
    Supplier Number: 229281
    Specifications:
    New Product
    New Product
  • Bulk Packed, tab lock box, non-sterile
    9"
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $287.56
    $287.56
    Case of 1,000
       $287.56
    Supplier Number: 229284
    Specifications:
    New Product
    New Product
  • Bulk packed, sterile
    9"
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $259.12
    $259.12
    Case of 500
       $259.12
    Supplier Number: 229286
    Specifications:
    New Product
    New Product
  • Bulk packed, non-sterile
    9"
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $197.50
    $197.50
    Case of 500
       $197.50
    Supplier Number: 229287
    Specifications:
    New Product
    New Product
Aspirating Bulb
Product Information
  • Désignation
    Longueur
    Availability
    Price
  • Rubber aspirating bulb for Plasteur® pipets, non-sterile
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $53.72
    $53.72
    Case of 12
       $53.72
    Supplier Number: 229279
    Specifications:
    New Product
    New Product