Plasteur® Pasteur Pipets
New Product
Supplier: CELLTREAT Scientific Products
Jump to:
About this item
Plasteur pipets for transferring liquids.
- CELLTREAT® exclusive product
Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
Flexible plastic helps prevent glove or skin punctures
Available packaged sterile individually wrapped or bulk packed
Available packaged non-sterile in re-sealable bags or tab lock boxes
Eliminate sharps disposal costs
13 Options Available Below
Product Details & Documents
Plasteur pipets for transferring liquids.
- CELLTREAT® exclusive product
Polypropylene is autoclavable and chemical resistant and provides chemical resistivity needed for many applications
Flexible plastic helps prevent glove or skin punctures
Available packaged sterile individually wrapped or bulk packed
Available packaged non-sterile in re-sealable bags or tab lock boxes
Eliminate sharps disposal costs
Product Family Options
Polystyrene Plasteur® Pipets
Product Information
- DésignationLongueurAvailabilityPrice
- Supplier Number: 229270Specifications:New ProductNew Product
- Supplier Number: 229275Specifications:New ProductNew Product
- Supplier Number: 229280Specifications:New ProductNew Product
- Supplier Number: 229285Specifications:New ProductNew Product
Polypropylene Plasteur® Pipets
Product Information
- DésignationLongueurAvailabilityPrice
- Supplier Number: 229271Specifications:New ProductNew Product
- Supplier Number: 229274Specifications:New ProductNew Product
- Supplier Number: 229276Specifications:New ProductNew Product
- Supplier Number: 229277Specifications:New ProductNew Product
- Supplier Number: 229281Specifications:New ProductNew Product
- Supplier Number: 229284Specifications:New ProductNew Product
- Supplier Number: 229286Specifications:New ProductNew Product
- Supplier Number: 229287Specifications:New ProductNew Product
Aspirating Bulb
Product Information
- DésignationLongueurAvailabilityPrice
- Supplier Number: 229279Specifications:New ProductNew Product