Solution Bottles, Sterile | Avantor
Solution Bottles, Sterile
New Product
Solution Bottles, Sterile
New Product
Certificates
Solution Bottles, Sterile
Solution Bottles, Sterile
4 Options Available Below
Product Details & Documents
Solution bottles are key for storage, preparation, and dispensing of chemical solutions and reagents.
  • Compatible with bottle top filters or for use as solution storage containers
    Solution bottle neck size is 45 mm
    Curved bottle design for easy handling
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Matière
    Color
    Availability
    Price
  • Solution bottle, sterile
    150 ml
    Polystyrene
    Clear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $159.52
    $159.52
    Case of 24
       $159.52
    Supplier Number: 229781
    Spécifications:
    New Product
    New Product
  • Solution bottle, sterile
    250 ml
    Polystyrene
    Clear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $172.75
    $172.75
    Case of 24
       $172.75
    Supplier Number: 229782
    Spécifications:
    New Product
    New Product
  • Solution bottle, sterile
    500 ml
    Polystyrene
    Clear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $238.34
    $238.34
    Case of 24
       $238.34
    Supplier Number: 229784
    Spécifications:
    New Product
    New Product
  • Solution bottle, sterile
    1000 ml
    Polystyrene
    Clear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $404.90
    $404.90
    Case of 24
       $404.90
    Supplier Number: 229785
    Spécifications:
    New Product