Multi-Function Laboratory Timer
Multi-function timer, 6×9×7"
Catalog # 77973-346
New Product
Supplier: CELLTREAT Scientific Products
Fournisseur #: 230123
Multi-Function Laboratory Timer
Catalog # 77973-346
Supplier: CELLTREAT Scientific Products
New Product
Jump to:
Product Details & Documents
Multi-function lab timers for accurate and efficient research to time multiple events simultaneously, track elapsed time, and manage time-sensitive procedures.
- Multiple functions include timer, stopwatch, clock, and alarm
- Easel stand, mounting magnet, and retractable hook for hanging
- White finish
- Uses one standard AAA battery (not included)
Improve the accuracy and reproducibility of experiments, especially when dealing with strict time schedules.
Spécifications
- Désignation:Multi-function timer
- Dimensions:6×9×7"