Replogle Globes Canada Wall Map
Canada wall map, 33", 49"
470360-132
New Product
: 72117
This Canadian map shows individual provincial flags and boundaries.
Folded map
- Wall mountable
- Paper construction
A Canadian wall map is a valuable educational tool that helps students visualize the country's geography, political boundaries, and regional diversity. It supports spatial learning and enhances understanding of Canada's physical and cultural landscape. Ideal for classrooms, it encourages curiosity and geographic literacy.
- Largeur:33"
- Désignation:Canada wall map
- Hauteur:49"