Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Fixation Buffer (Flow Cytometry), Abcam | Avantor

Fixation Buffer (Flow Cytometry), Abcam

Fixation Buffer (Flow Cytometry), 100 ml
Catalog # ABCA_AB314680-100M
New Product
Supplier:   Abcam
Fournisseur #:  AB314680-100ML
Fixation Buffer (Flow Cytometry), Abcam
Catalog # ABCA_AB314680-100M
Supplier:  Abcam
New Product
CAS Number:  
Fixation Buffer (Flow Cytometry), Abcam

Fixation Buffer (Flow Cytometry), 100 ml

CAS Number:  
Price
$400.00
Log in pour la disponibilité
Each (100ml)
La quantité minimale de commande est de 1
Product Details & Documents

Fixation Buffer (ab314680) is a ready-to-use formaldehyde-based fixation buffer for immunofluorescence cell staining for microscopy or flow cytometry.

  • pH: 7.4 Constituents: 5% Formaldehyde

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Désignation:
    Fixation Buffer (Flow Cytometry)
  • Taille:
    100 ml
  • Cat. No.:
    ABCA_AB314680-100M
  • Storage temperature:
    Store at Ambient.

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.