Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam | Avantor

Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam

New Product
Supplier:   Abcam
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam
Supplier:  Abcam
New Product
Certificates
Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam

2 Options Available Below

Product Details & Documents

Tissue Clearing Permeabilization Buffer ab243307 is part of our range of tissue clearing reagents and products that enable you to set up tissue clearing easily and quickly.

  • Constituents: 94% Water, 6% Sodium lauryl sulfate

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications for Product Family

Spécifications

Spécifications

  • Catalog No:
  • ABCA_AB243307-30ML
  • ABCA_AB243307-100M
  • Storage temperature:
  • Store at Ambient.
  • Store at Ambient.

Spécifications

Product Family Options

Informations sur le produit

  • Désignation
    Taille
    Availability
    Price
  • Tissue Clearing Permeabilization Buffer - hydrophobic
    30 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (30ml)
    Each (30ml) - $195.00
    $195.00
    Each (30ml)
       $195.00
    La quantité minimale de commande est de 1

    Supplier Number: AB243307-30ML
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB243307-30ML
    Storage temperature
    Store at Ambient.
    New Product
  • Tissue Clearing Permeabilization Buffer - hydrophobic
    100 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (100ml)
    Each (100ml) - $359.00
    $359.00
    Each (100ml)
       $359.00
    La quantité minimale de commande est de 1

    Supplier Number: AB243307-100ML
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCA_AB243307-100M
    Storage temperature
    Store at Ambient.
    New Product