Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
ChIP Buffer, Abcam

ChIP Buffer, Abcam

Catalog # ABCA_AB205698-6ML
New Product
Supplier:   Abcam
Supplier #:  AB205698-6ML
ChIP Buffer, Abcam
Catalog # ABCA_AB205698-6ML
Supplier:  Abcam
New Product
CAS Number:  
ChIP Buffer, Abcam
CAS Number:  
Price
$257.00
Log in pour la disponibilité
Each (6ml)
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

ChIP Buffer for ab185913

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spécifications

Spécifications

  • Cat. No.:
    ABCA_AB205698-6ML
  • Storage temperature:
    Store at +4°C.

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.