Native lysis Buffer, Abcam
Native lysis Buffer, 15 ml
ABCA_AB156035-15ML
New Product
: Abcam
: AB156035-15ML
Abcam’s 1X Native lysis buffer is an efficient means of cell lysis and protein solubilization for both adherent and suspension cultured mammalian cells.
- pH: 7.5 Constituents: 1.5% Dodecyl maltoside, 0.6% HEPES, 0.58% Sodium chloride, 0.11% Sodium pyrophosphate decahydrate, 0.04% EGTA, 0.03% EDTA, 0.02% Sodium orthovanadate, 0.02% disodium;1,3-dihydroxypropan-2-yl phosphate;hydrate
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Désignation:Native lysis Buffer
- Taille:15 ml
- Cat. No.:ABCA_AB156035-15ML
- Storage temperature:Store at +4°C.



