Eosin Y solution modifiée en solution alcoolique | Avantor
Eosin Y solution modifiée en solution alcoolique
500 ml
Catalog # ABCA_AB246824-500M
New Product
Supplier:   Abcam
Fournisseur #:  AB246824-500ML
Eosin Y solution modifiée en solution alcoolique
Catalog # ABCA_AB246824-500M
Supplier:  Abcam
New Product
CAS Number:  
Eosin Y solution modifiée en solution alcoolique
500 ml
CAS Number:  
Price
$287.00
Log in pour la disponibilité
Each (500ml)
La quantité minimale de commande est de 1
Product Details & Documents
Eosin Y solution (modified alcoholic) is a histology and cytology stain offering improved usability for tissue section staining. - Improved usability – lower evaporation rate and enhanced surface tension - Color patterns – stains cytoplasm light pink, collagen pink, muscle pink/rose, and erythrocytes pink/red - Control tissue – suitable for paraffin-embedded, frozen tissue sections, and cell smears
  • Constituents: 65% Ethanol, 3.8% Isopropanol, 1% Acetic acid, 0.25% Eosin Y
 : All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spécifications
Spécifications
  • Taille:
    500 ml
Spécifications