Eosin Y solution en solution alcoolique | Avantor
Eosin Y solution en solution alcoolique
500 ml
Catalog # ABCA_AB246823-500M
New Product
Supplier:   Abcam
Fournisseur #:  AB246823-500ML
Eosin Y solution en solution alcoolique
Catalog # ABCA_AB246823-500M
Supplier:  Abcam
New Product
CAS Number:  
Eosin Y solution en solution alcoolique
500 ml
CAS Number:  
Price
$287.00
Log in pour la disponibilité
Each (500ml)
La quantité minimale de commande est de 1
Product Details & Documents
Eosin Y Solution (Alcoholic) ab246823 is intended for use in the histological demonstration of cytoplasm and is commonly used as a counterstain for Hematoxylin.
  • pH: 3 - 5 Constituents: 80% Ethanol, 3.8% Isopropanol, 1% Acetic acid, 0.25% Eosin Y
Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spécifications
Spécifications
  • Taille:
    500 ml
Spécifications