Congo red solution
500 ml
ABCA_AB246819-500M
New Product
: Abcam
: AB246819-500ML
Congo Red Solution ab246819 is intended for use in the histological visualization of amyloid in tissue sections.
- pH: 9 - 11 Constituents: 48% Ethanol, 2% Isopropanol, 0.5% Congo Red
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Taille:500 ml