Proteometer-CV Fluorescent Reagent
Proteometer-CV Fluorescent Reagent Vial (1X), to be stored at -20 °C
  78047-218
New Product
 :  RGN-PCV-001
Proteometer-CV Fluorescent Reagent
  78047-218
New Product
 :  
Proteometer-CV Fluorescent Reagent
Proteometer-CV Fluorescent Reagent
Proteometer-CV Fluorescent Reagent Vial (1X), to be stored at -20 °C
 :  
$2,103.30
Each
1 vial of Fluorescent reagent for the Proteometer-CV kit specifically formulated for use with Proteometer-CV kit.
  • Vials of the Proteometer-CV fluorescent reagent for reorder to continue analysis with the Proteometer-CV kit
Intended for use with the Proteometer-CV kit.
 : Reorder individually as needed.
 : Supplied in a custom box with instructions for use.
  • Désignation:
    Proteometer-CV Fluorescent Reagent Vial (1X), to be stored at -20 °C