Proteometer-CV Fluorescent Reagent
Proteometer-CV Fluorescent Reagent Vial (1X), to be stored at -20 °C
: RGN-PCV-001
1 vial of Fluorescent reagent for the Proteometer-CV kit specifically formulated for use with Proteometer-CV kit.
Vials of the Proteometer-CV fluorescent reagent for reorder to continue analysis with the Proteometer-CV kit
Intended for use with the Proteometer-CV kit.
: Reorder individually as needed.
: Supplied in a custom box with instructions for use.
- Désignation:Proteometer-CV Fluorescent Reagent Vial (1X), to be stored at -20 °C