Proteometer-CV Reactor
Proteometer-CV Reactor
Catalog # 78047-214
New Product
Supplier: NOVILYTIC, LLC
Supplier #: RCT-PCV-001
Jump to:
Product Details & Documents
Proteometer-CV Reactor sold individually.
Proteometer-CV Reactor replaces the LC column in the analytical system
Intended for use with the Proteometer-CV kit.
Ordering information: Reorder individually as needed.
Packaging: Supplied in a custom box with instructions for use. Follow instructions in "Instructions for Use" for proper storage.
Specifications
- Désignation:Proteometer-CV Reactor