Proteometer-CV Reactor
Proteometer-CV Reactor
Proteometer-CV Reactor
Catalog # 78047-214
New Product
Supplier:   NOVILYTIC, LLC
Supplier #:  RCT-PCV-001
Proteometer-CV Reactor
Catalog # 78047-214
Supplier:  NOVILYTIC, LLC
New Product
Proteometer-CV Reactor
Proteometer-CV Reactor
Proteometer-CV Reactor
Price
$4,530.18
Log in for availability
Each
Product Details & Documents
Proteometer-CV Reactor sold individually.
  • Proteometer-CV Reactor replaces the LC column in the analytical system
Intended for use with the Proteometer-CV kit.
Ordering information: Reorder individually as needed.
Packaging: Supplied in a custom box with instructions for use. Follow instructions in "Instructions for Use" for proper storage.
Specifications
  • Désignation:
    Proteometer-CV Reactor