VWR® Filter Papers
VWR® filter papers are high-performance cellulose-based filtration materials designed for accurate and reproducible separations in laboratory settings. Available in both qualitative and quantitative grades, these papers deliver consistent retention, flow rate, and purity across a wide range of scientific applications. Whether used in gravimetric analysis, sample cleanup, or routine laboratory workflows, VWR® filter papers provide the reliability and technical precision demanded by today's researchers.
Manufactured from high-purity cellulose fibers for low background interference
Available in qualitative (grades 410–417) and quantitative (grade 494) formats
Multiple grades, diameters, and porosity types to suit specific lab workflows
Optimized for consistent particle retention and flow rate performance
Low ash content options ideal for gravimetric and residue-sensitive testing
Compatible with a wide range of funnel systems and filtration setups
VWR® filter papers are engineered to meet the demanding needs of laboratories across industries including environmental testing, pharmaceutical analysis, food and beverage quality control, and academic research. These papers are available in a range of pore sizes and porosity profiles to suit everything from coarse precipitate separations to fine particulate capture. Qualitative grades such as 410, 413, and 454 are ideal for routine separations and clarity-based filtration, while quantitative grades like 494 are ashless and optimized for precision gravimetric work. All grades are processed under strict quality standards, ensuring uniform thickness, excellent wet strength, and reliable filtration characteristics. With a broad range of diameters, formats, and retention levels, VWR® filter papers integrate easily into existing lab workflows and help ensure repeatable, high-quality analytical results.
VWR® filter papers are high-performance cellulose-based filtration materials designed for accurate and reproducible separations in laboratory settings. Available in both qualitative and quantitative grades, these papers deliver consistent retention, flow rate, and purity across a wide range of scientific applications. Whether used in gravimetric analysis, sample cleanup, or routine laboratory workflows, VWR® filter papers provide the reliability and technical precision demanded by today's researchers.
Manufactured from high-purity cellulose fibers for low background interference
Available in qualitative (grades 410–417) and quantitative (grade 494) formats
Multiple grades, diameters, and porosity types to suit specific lab workflows
Optimized for consistent particle retention and flow rate performance
Low ash content options ideal for gravimetric and residue-sensitive testing
Compatible with a wide range of funnel systems and filtration setups
VWR® filter papers are engineered to meet the demanding needs of laboratories across industries including environmental testing, pharmaceutical analysis, food and beverage quality control, and academic research. These papers are available in a range of pore sizes and porosity profiles to suit everything from coarse precipitate separations to fine particulate capture. Qualitative grades such as 410, 413, and 454 are ideal for routine separations and clarity-based filtration, while quantitative grades like 494 are ashless and optimized for precision gravimetric work. All grades are processed under strict quality standards, ensuring uniform thickness, excellent wet strength, and reliable filtration characteristics. With a broad range of diameters, formats, and retention levels, VWR® filter papers integrate easily into existing lab workflows and help ensure repeatable, high-quality analytical results.
Product Family Options
Grade 454 – General-Purpose Qualitative Filter Paper with Medium Retention
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-454New Product - New Product
Specifications
Cat. No.77969-456New Product - New Product
Specifications
Cat. No.77969-458New Product - New Product
Specifications
Cat. No.77969-460New Product - New Product
Specifications
Cat. No.77969-462New Product - New Product
Specifications
Cat. No.77969-464New Product
Grade 474 – Medium Qualitative Filter Paper with Slightly Slower Flow
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-472New Product - New Product
Specifications
Cat. No.77969-466New Product - New Product
Specifications
Cat. No.77969-468New Product - New Product
Specifications
Cat. No.77969-470New Product
Grade 494 – Fine Quantitative Filter Paper with Slow Flow Rate
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-474New Product - New Product
Specifications
Cat. No.77969-452New Product
Grade 313 - Thick, Durable Filter Paper for Bulk Precipitate Filtration
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-510New Product - New Product
Specifications
Cat. No.77969-512New Product - New Product
Specifications
Cat. No.77969-514New Product - New Product
Specifications
Cat. No.77969-516New Product - New Product
Specifications
Cat. No.77969-518New Product - New Product
Specifications
Cat. No.77969-520New Product
Grade 315 – Crepe Fluted Paper for Coarse and Gelatinous Precipitates
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-532New Product - New Product
Specifications
Cat. No.77969-522New Product - New Product
Specifications
Cat. No.77969-524New Product - New Product
Specifications
Cat. No.77969-526New Product - New Product
Specifications
Cat. No.77969-528New Product - New Product
Specifications
Cat. No.77969-530New Product
Grade 410 – Fine Qualitative Filter Paper with Slow Flow Rate
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-534New Product - New Product
Specifications
Cat. No.77969-536New Product - New Product
Specifications
Cat. No.77969-538New Product - New Product
Specifications
Cat. No.77969-540New Product
Grade 413 – Medium Qualitative Filter Paper for General Laboratory Use
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-542New Product - New Product
Specifications
Cat. No.77969-544New Product - New Product
Specifications
Cat. No.77969-546New Product - New Product
Specifications
Cat. No.77969-548New Product - New Product
Specifications
Cat. No.77969-550New Product - New Product
Specifications
Cat. No.77969-552New Product - New Product
Specifications
Cat. No.77969-554New Product - New Product
Specifications
Cat. No.77969-556New Product - New Product
Specifications
Cat. No.77969-558New Product - New Product
Specifications
Cat. No.77969-560New Product - New Product
Specifications
Cat. No.77969-562New Product - New Product
Specifications
Cat. No.77969-564New Product - New Product
Specifications
Cat. No.77969-566New Product - New Product
Specifications
Cat. No.77969-568New Product
Grade 415 – Coarse Qualitative Filter Paper for Rapid Filtration
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-570New Product - New Product
Specifications
Cat. No.77969-572New Product - New Product
Specifications
Cat. No.77969-574New Product - New Product
Specifications
Cat. No.77969-576New Product - New Product
Specifications
Cat. No.77969-578New Product - New Product
Specifications
Cat. No.77969-580New Product - New Product
Specifications
Cat. No.77969-582New Product - New Product
Specifications
Cat. No.77969-584New Product - New Product
Specifications
Cat. No.77969-586New Product - New Product
Specifications
Cat. No.77969-588New Product - New Product
Specifications
Cat. No.77969-590New Product
Grade 417 - Very coarse, high-speed filter paper
- GradeØDébitParticle RetentionTaille de pore
- New Product
Specifications
Cat. No.77969-592New Product - New Product
Specifications
Cat. No.77969-612New Product - New Product
Specifications
Cat. No.77969-614New Product - New Product
Specifications
Cat. No.77969-594New Product - New Product
Specifications
Cat. No.77969-596New Product - New Product
Specifications
Cat. No.77969-598New Product - New Product
Specifications
Cat. No.77969-600New Product - New Product
Specifications
Cat. No.77969-602New Product - New Product
Specifications
Cat. No.77969-604New Product - New Product
Specifications
Cat. No.77969-606New Product - New Product
Specifications
Cat. No.77969-608New Product - New Product
Specifications
Cat. No.77969-610New Product