VWR® Filter Papers
New Product
VWR® Filter Papers
New Product
VWR® Filter Papers
VWR® Filter Papers
VWR® filter papers are high-performance cellulose-based filtration materials designed for accurate and reproducible separations in laboratory settings. Available in both qualitative and quantitative grades, these papers deliver consistent retention, flow rate, and purity across a wide range of scientific applications. Whether used in gravimetric analysis, sample cleanup, or routine laboratory workflows, VWR® filter papers provide the reliability and technical precision demanded by today's researchers.
  • Manufactured from high-purity cellulose fibers for low background interference
    Available in qualitative (grades 410–417) and quantitative (grade 494) formats
    Multiple grades, diameters, and porosity types to suit specific lab workflows
    Optimized for consistent particle retention and flow rate performance
    Low ash content options ideal for gravimetric and residue-sensitive testing
    Compatible with a wide range of funnel systems and filtration setups
  •  
VWR® filter papers are engineered to meet the demanding needs of laboratories across industries including environmental testing, pharmaceutical analysis, food and beverage quality control, and academic research. These papers are available in a range of pore sizes and porosity profiles to suit everything from coarse precipitate separations to fine particulate capture. Qualitative grades such as 410, 413, and 454 are ideal for routine separations and clarity-based filtration, while quantitative grades like 494 are ashless and optimized for precision gravimetric work. All grades are processed under strict quality standards, ensuring uniform thickness, excellent wet strength, and reliable filtration characteristics. With a broad range of diameters, formats, and retention levels, VWR® filter papers integrate easily into existing lab workflows and help ensure repeatable, high-quality analytical results.
Product Family Options
Grade 454 – General-Purpose Qualitative Filter Paper with Medium Retention
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 454
    5.5 cm
    Medium-Fast
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 29,23 $
    29,23 $
    Pack of 100
       29,23 $
    New Product
    Specifications
    Cat. No.
    77969-454
    New Product
  • 454
    9 cm
    Medium-Fast
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 47,79 $
    47,79 $
    Pack of 100
       47,79 $
    New Product
    Specifications
    Cat. No.
    77969-456
    New Product
  • 454
    11 cm
    Medium-Fast
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 60,22 $
    60,22 $
    Pack of 100
       60,22 $
    New Product
    Specifications
    Cat. No.
    77969-458
    New Product
  • 454
    12.5 cm
    Medium-Fast
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 68,47 $
    68,47 $
    Pack of 100
       68,47 $
    New Product
    Specifications
    Cat. No.
    77969-460
    New Product
  • 454
    15 cm
    Medium-Fast
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 82,10 $
    82,10 $
    Pack of 100
       82,10 $
    New Product
    Specifications
    Cat. No.
    77969-462
    New Product
  • 454
    18.5 cm
    Medium-Fast
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 101,26 $
    101,26 $
    Pack of 100
       101,26 $
    New Product
    Specifications
    Cat. No.
    77969-464
    New Product
Grade 474 – Medium Qualitative Filter Paper with Slightly Slower Flow
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 474
    15 cm
    Medium-Slow
    5 - 10 µm
    Medium
    Pack of 100
    Pack of 100 - 82,05 $
    82,05 $
    Pack of 100
       82,05 $
    New Product
    Specifications
    Cat. No.
    77969-472
    New Product
  • 474
    9 cm
    Medium-Slow
    5 - 10 µm
    Medium
    Pack of 100
    Pack of 100 - 49,31 $
    49,31 $
    Pack of 100
       49,31 $
    New Product
    Specifications
    Cat. No.
    77969-466
    New Product
  • 474
    11 cm
    Medium-Slow
    5 - 10 µm
    Medium
    Pack of 100
    Pack of 100 - 60,28 $
    60,28 $
    Pack of 100
       60,28 $
    New Product
    Specifications
    Cat. No.
    77969-468
    New Product
  • 474
    12.5 cm
    Medium-Slow
    5 - 10 µm
    Medium
    Pack of 100
    Pack of 100 - 68,50 $
    68,50 $
    Pack of 100
       68,50 $
    New Product
    Specifications
    Cat. No.
    77969-470
    New Product
Grade 494 – Fine Quantitative Filter Paper with Slow Flow Rate
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 494
    11 cm
    Slow
    1 µm
    Fine
    Pack of 100
    Pack of 100 - 72,61 $
    72,61 $
    Pack of 100
       72,61 $
    New Product
    Specifications
    Cat. No.
    77969-474
    New Product
  • 494
    12.5 cm
    Slow
    1 µm
    Fine
    Pack of 100
    Pack of 100 - 72,64 $
    72,64 $
    Pack of 100
       72,64 $
    New Product
    Specifications
    Cat. No.
    77969-452
    New Product
Grade 313 - Thick, Durable Filter Paper for Bulk Precipitate Filtration
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 313
    12.5 cm
    Medium-Fast
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 45,63 $
    45,63 $
    Pack of 100
       45,63 $
    New Product
    Specifications
    Cat. No.
    77969-510
    New Product
  • 313
    15 cm
    Medium
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 54,77 $
    54,77 $
    Pack of 100
       54,77 $
    New Product
    Specifications
    Cat. No.
    77969-512
    New Product
  • 313
    18.5 cm
    Medium
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 67,52 $
    67,52 $
    Pack of 100
       67,52 $
    New Product
    Specifications
    Cat. No.
    77969-514
    New Product
  • 313
    24 cm
    Medium
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 87,57 $
    87,57 $
    Pack of 100
       87,57 $
    New Product
    Specifications
    Cat. No.
    77969-516
    New Product
  • 313
    32 cm
    Medium
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 116,80 $
    116,80 $
    Pack of 100
       116,80 $
    New Product
    Specifications
    Cat. No.
    77969-518
    New Product
  • 313
    38.5 cm
    Medium
    10 µm
    Medium
    Pack of 100
    Pack of 100 - 140,53 $
    140,53 $
    Pack of 100
       140,53 $
    New Product
    Specifications
    Cat. No.
    77969-520
    New Product
Grade 315 – Crepe Fluted Paper for Coarse and Gelatinous Precipitates
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 315
    38.5 cm
    Fast
    25 µm
    Course
    Pack of 100
    Pack of 100 - 134,11 $
    134,11 $
    Pack of 100
       134,11 $
    New Product
    Specifications
    Cat. No.
    77969-532
    New Product
  • 315
    12.5 cm
    Fast
    25 µm
    Course
    Pack of 100
    Pack of 100 - 43,55 $
    43,55 $
    Pack of 100
       43,55 $
    New Product
    Specifications
    Cat. No.
    77969-522
    New Product
  • 315
    15 cm
    Fast
    25 µm
    Course
    Pack of 100
    Pack of 100 - 52,25 $
    52,25 $
    Pack of 100
       52,25 $
    New Product
    Specifications
    Cat. No.
    77969-524
    New Product
  • 315
    18.5 cm
    Fast
    25 µm
    Course
    Pack of 100
    Pack of 100 - 64,42 $
    64,42 $
    Pack of 100
       64,42 $
    New Product
    Specifications
    Cat. No.
    77969-526
    New Product
  • 315
    24 cm
    Fast
    25 µm
    Course
    Pack of 100
    Pack of 100 - 83,63 $
    83,63 $
    Pack of 100
       83,63 $
    New Product
    Specifications
    Cat. No.
    77969-528
    New Product
  • 315
    32 cm
    Fast
    25 µm
    Course
    Pack of 10
    Pack of 10 - 111,51 $
    111,51 $
    Pack of 10
       111,51 $
    New Product
    Specifications
    Cat. No.
    77969-530
    New Product
Grade 410 – Fine Qualitative Filter Paper with Slow Flow Rate
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 410
    9 cm
    Slow
    1 µm
    Fine
    Pack of 100
    Pack of 100 - 35,78 $
    35,78 $
    Pack of 100
       35,78 $
    New Product
    Specifications
    Cat. No.
    77969-534
    New Product
  • 410
    12.5 cm
    Slow
    1 µm
    Fine
    Pack of 100
    Pack of 100 - 49,81 $
    49,81 $
    Pack of 100
       49,81 $
    New Product
    Specifications
    Cat. No.
    77969-536
    New Product
  • 410
    15 cm
    Slow
    1 µm
    Fine
    Pack of 100
    Pack of 100 - 59,78 $
    59,78 $
    Pack of 100
       59,78 $
    New Product
    Specifications
    Cat. No.
    77969-538
    New Product
  • 410
    18.5 cm
    Slow
    1 µm
    Fine
    Pack of 100
    Pack of 100 - 73,67 $
    73,67 $
    Pack of 100
       73,67 $
    New Product
    Specifications
    Cat. No.
    77969-540
    New Product
Grade 413 – Medium Qualitative Filter Paper for General Laboratory Use
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 413
    1.5 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 34,50 $
    34,50 $
    Pack of 100
       34,50 $
    New Product
    Specifications
    Cat. No.
    77969-542
    New Product
  • 413
    4.25 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 18,30 $
    18,30 $
    Pack of 100
       18,30 $
    New Product
    Specifications
    Cat. No.
    77969-544
    New Product
  • 413
    5.5 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 23,72 $
    23,72 $
    Pack of 100
       23,72 $
    New Product
    Specifications
    Cat. No.
    77969-546
    New Product
  • 413
    7.5 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 32,35 $
    32,35 $
    Pack of 100
       32,35 $
    New Product
    Specifications
    Cat. No.
    77969-548
    New Product
  • 413
    9 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 38,81 $
    38,81 $
    Pack of 100
       38,81 $
    New Product
    Specifications
    Cat. No.
    77969-550
    New Product
  • 413
    11 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 47,38 $
    47,38 $
    Pack of 100
       47,38 $
    New Product
    Specifications
    Cat. No.
    77969-552
    New Product
  • 413
    12.5 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 53,92 $
    53,92 $
    Pack of 100
       53,92 $
    New Product
    Specifications
    Cat. No.
    77969-554
    New Product
  • 413
    15 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 64,69 $
    64,69 $
    Pack of 100
       64,69 $
    New Product
    Specifications
    Cat. No.
    77969-556
    New Product
  • 413
    18.5 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 79,79 $
    79,79 $
    Pack of 100
       79,79 $
    New Product
    Specifications
    Cat. No.
    77969-558
    New Product
  • 413
    20.5 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 88,51 $
    88,51 $
    Pack of 100
       88,51 $
    New Product
    Specifications
    Cat. No.
    77969-560
    New Product
  • 413
    25 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 107,78 $
    107,78 $
    Pack of 100
       107,78 $
    New Product
    Specifications
    Cat. No.
    77969-562
    New Product
  • 413
    33 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 142,34 $
    142,34 $
    Pack of 100
       142,34 $
    New Product
    Specifications
    Cat. No.
    77969-564
    New Product
  • 413
    38.5 cm
    Medium
    5 µm
    Medium
    Pack of 100
    Pack of 100 - 166,07 $
    166,07 $
    Pack of 100
       166,07 $
    New Product
    Specifications
    Cat. No.
    77969-566
    New Product
  • 413
    20X20 I
    Medium
    5 µm
    Medium
    Pack of 50
    Pack of 50 - 92,89 $
    92,89 $
    Pack of 50
       92,89 $
    New Product
    Specifications
    Cat. No.
    77969-568
    New Product
Grade 415 – Coarse Qualitative Filter Paper for Rapid Filtration
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 415
    5.5 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 25,50 $
    25,50 $
    Pack of 100
       25,50 $
    New Product
    Specifications
    Cat. No.
    77969-570
    New Product
  • 415
    7.5 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 34,85 $
    34,85 $
    Pack of 100
       34,85 $
    New Product
    Specifications
    Cat. No.
    77969-572
    New Product
  • 415
    9 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 41,71 $
    41,71 $
    Pack of 100
       41,71 $
    New Product
    Specifications
    Cat. No.
    77969-574
    New Product
  • 415
    11 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 51,06 $
    51,06 $
    Pack of 100
       51,06 $
    New Product
    Specifications
    Cat. No.
    77969-576
    New Product
  • 415
    12.5 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 58,02 $
    58,02 $
    Pack of 100
       58,02 $
    New Product
    Specifications
    Cat. No.
    77969-578
    New Product
  • 415
    15 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 69,71 $
    69,71 $
    Pack of 100
       69,71 $
    New Product
    Specifications
    Cat. No.
    77969-580
    New Product
  • 415
    18.5 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 85,94 $
    85,94 $
    Pack of 100
       85,94 $
    New Product
    Specifications
    Cat. No.
    77969-582
    New Product
  • 415
    20.5 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 95,28 $
    95,28 $
    Pack of 100
       95,28 $
    New Product
    Specifications
    Cat. No.
    77969-584
    New Product
  • 415
    25 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 116,17 $
    116,17 $
    Pack of 100
       116,17 $
    New Product
    Specifications
    Cat. No.
    77969-586
    New Product
  • 415
    33 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 153,28 $
    153,28 $
    Pack of 100
       153,28 $
    New Product
    Specifications
    Cat. No.
    77969-588
    New Product
  • 415
    38.5 cm
    Fast
    25 µm
    Coarse
    Pack of 100
    Pack of 100 - 178,85 $
    178,85 $
    Pack of 100
       178,85 $
    New Product
    Specifications
    Cat. No.
    77969-590
    New Product
Grade 417 - Very coarse, high-speed filter paper
  • Grade
    Ø
    Débit
    Particle Retention
    Taille de pore
  • 417
    5.5 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 27,38 $
    27,38 $
    Pack of 50
       27,38 $
    New Product
    Specifications
    Cat. No.
    77969-592
    New Product
  • 417
    40 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 199,16 $
    199,16 $
    Pack of 50
       199,16 $
    New Product
    Specifications
    Cat. No.
    77969-612
    New Product
  • 417
    50 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 248,95 $
    248,95 $
    Pack of 50
       248,95 $
    New Product
    Specifications
    Cat. No.
    77969-614
    New Product
  • 417
    7.5 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 37,34 $
    37,34 $
    Pack of 50
       37,34 $
    New Product
    Specifications
    Cat. No.
    77969-594
    New Product
  • 417
    9 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 44,82 $
    44,82 $
    Pack of 50
       44,82 $
    New Product
    Specifications
    Cat. No.
    77969-596
    New Product
  • 417
    11 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 54,75 $
    54,75 $
    Pack of 50
       54,75 $
    New Product
    Specifications
    Cat. No.
    77969-598
    New Product
  • 417
    12.5 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 62,19 $
    62,19 $
    Pack of 50
       62,19 $
    New Product
    Specifications
    Cat. No.
    77969-600
    New Product
  • 417
    15 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 74,81 $
    74,81 $
    Pack of 50
       74,81 $
    New Product
    Specifications
    Cat. No.
    77969-602
    New Product
  • 417
    18.5 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 92,05 $
    92,05 $
    Pack of 50
       92,05 $
    New Product
    Specifications
    Cat. No.
    77969-604
    New Product
  • 417
    20.5 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 102,11 $
    102,11 $
    Pack of 50
       102,11 $
    New Product
    Specifications
    Cat. No.
    77969-606
    New Product
  • 417
    25 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 124,43 $
    124,43 $
    Pack of 50
       124,43 $
    New Product
    Specifications
    Cat. No.
    77969-608
    New Product
  • 417
    33 cm
    Fast
    40 µm
    Coarse
    Pack of 50
    Pack of 50 - 164,24 $
    164,24 $
    Pack of 50
       164,24 $
    New Product
    Specifications
    Cat. No.
    77969-610