Proteometer-CV Consumables Kit
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Proteometer-CV is designed for the scientific development of mAbs, bi-specifics, or fusion proteins with a human Fc region, regardless of their stage in the development continuum. This kit offers an alternative to traditional charge variant methods that require Protein A.
No Sample Preparation Required - analysis directly from clarified fermentation broth without Protein A purification
- Speed and Efficiency -complete your CV analysis in under 25 minutes
- Simplified Workflow - all the components you need for seamless operation
- Exclusive Technology - the only solution that performs charge variant analysis directly in clarified fermentation broth
Determination of mAb charge variants and titer of therapeutic mAbs and other proteins that contain the Fc domain of human IgG. It is intended primarily to monitor mAb quality directly in clarified fermentation broth without Protein A purification during product development. The method utilizes HPLC systems equipped with two pumps and a fluorescence detector.
Accessories information: See the Proteometer-CV Reactor, Proteometer-CV Buffer Packets, and Proteometer-CV Reagent series for accessory component replacement options.
Delivery information:
Packaging: Supplied in a custom box with instructions for use.
Specifications
Specifications
- Désignation:Proteometer-CV consumables kit, reagent to be stored at -20 °C