G-Biosciences TBE Running Buffer (10X)
Catalogue # CA82021-496
Fournisseur: G-Biosciences
Numéro CAS:
Spécifications
- Description:TBE (10X)
- Size:1 L
- Cat. no.:CA82021-496
- Supplier No.:R025
Spécifications
À propos de cet article
The primary use of TBE running buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
- 10X concentrated solution: 0.9 M tris-borate, 0.01 M EDTA (pH 8.3)
The buffer maintains consistent pH and conductivity, enabling sharp band resolution and protecting nucleic acids from degradation during electrophoresis.