ContactUsLinkComponent
G-Biosciences TBE Running Buffer (10X)
Catalogue # CA82021-496
Fournisseur:  G-Biosciences
Numéro CAS:  
G-Biosciences TBE Running Buffer (10X)
Catalogue # CA82021-496
Fournisseur:  G-Biosciences
Supplier Number:  R025
Numéro CAS:  
Spécifications
  • Description:
    TBE (10X)
  • Size:
    1 L
  • Cat. no.:
    CA82021-496
  • Supplier No.:
    R025
Spécifications
À propos de cet article
The primary use of TBE running buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
  • 10X concentrated solution: 0.9 M tris-borate, 0.01 M EDTA (pH 8.3)
The buffer maintains consistent pH and conductivity, enabling sharp band resolution and protecting nucleic acids from degradation during electrophoresis.