G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
  • 1 M Tris·HCl (pH 7.4)
  • 1.5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Product Family Options
  • Désignation
    Taille
  • Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl)
    1 L
    Each (1l)
    Each (1l) - $327.56
    $327.56
    Each (1l)
       $327.56
     : R017
  • Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl)
    1 gal.
    Each (4l)
    Each (4l) - $625.94
    $625.94
    Each (4l)
       $625.94
     : R018