G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
1 M Tris·HCl (pH 7.4)
- 1.5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
1 M Tris·HCl (pH 7.4)
- 1.5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Product Family Options
- DésignationTaille
- Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl)1 LEach (1l)$327.56: R017
- Hybridization neutralizing solution (1 M Tris.HCl (pH 7.4), 1.5 M NaCl)1 gal.Each (4l)$625.94: R018