Human Recombinant IL-18 (from E. coli)
Supplier: Sartorius
À propos de cet article
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
2 Options Available Below
Product Details & Documents
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 77870-807
- 77870-860
- Protein/peptide name:
- IL-18
- IL-18
- Protein synonyms:
- Interleukin-18
- Interleukin-18
- Protein/peptide type:
- Recombinant
- Recombinant
- Protein Function:
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Source:
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Protease-free:
- N
- N
- Animal-free:
- Y
- Y
- Nuclease-free:
- N
- N
- Carrier-free:
- Y
- Y
- Endotoxin-free:
- Y
- Y
- Storage conditions:
- Ambient
- Ambient
- Shipping temperature:
- Ambient
- Ambient
Spécifications
Product Family Options
Informations sur le produit
- Type de conditionnementTailleAvailabilityPrice
- Supplier Number: CYK-0025-1015Spécifications:
More
Specifications
Cat. No.77870-807Protein/peptide nameIL-18Protein synonymsInterleukin-18Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient - Supplier Number: CYK-0100-1015Spécifications:
More
Specifications
Cat. No.77870-860Protein/peptide nameIL-18Protein synonymsInterleukin-18Protein/peptide typeRecombinantProtein FunctionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedProtease-freeNAnimal-freeYNuclease-freeNCarrier-freeYEndotoxin-freeYStorage conditionsAmbientShipping temperatureAmbient
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



