NEBNext® Microbiome DNA Enrichment Kit
Supplier: New England Biolabs (NEB)
Certificates
About this item
The NEBNext Microbiome DNA enrichment kit facilitates enrichment of microbial DNA from samples containing methylated host DNA (including human), by selective binding and removal of the CpG-methylated host DNA. Importantly, microbial diversity remains intact after enrichment.
Effective enrichment of microbial genomic DNA from samples contaminating host DNA
- Fast, simple protocol
- Enables microbiome whole genome sequencing, even for samples with high levels of host DNA
- Compatible with downstream applications including next generation sequencing on all platforms, qPCR
- Suitable for a wide range of sample types
Frequently Bought Together