Estes Fin Alignment Guide
Fin alignment guide
Catalog # 470351-922
Supplier: Estes Industries
Fournisseur #: 2231
Jump to:
Product Details & Documents
Ensure perfect geometric alignment of your balsa fins on any Estes model rocket.
Fast and accurate fin alignment
- Stabilizes fins and body tube while glue sets
- Great for three or four finned rockets
- Fits body tubes ranging from BT-20 to BT-80
Never attach your fins wrong again and remove that painful process of holding them in place while the adhesive dries! A precision molded accessory that makes it easy to glue three or four fins to various sized body tubes ranging from BT-20 to BT-80 diameters. Unique and adjustable vertical fin holders can accommodate either ³/₃₂ or ¹/₈" balsa, bass or plywood fins. This is a must-have tool for any serious model rocket builder.
Spécifications
- Désignation:Fin alignment guide