About this item
These non-ionic detergents are suitable for isolating membrane-protein complexes.
G-Biosciences’ Proteomic Grade Detergent Solutions contain reduced peroxides and carbonyl compounds
In addition, the detergents have less than 50 µS conductivity
These detergents are offered as sterile, 10% aqueous solutions, sealed under inert gas, and are suitable for protein applications
Many commercial grade detergents contain elevated levels of sulfhydryl oxidizing agents, peroxides, salts and carbonyl compounds. The proteins that are isolated with these detergents are highly susceptible to contaminating peroxides and carbonyls. The peroxides will oxidize proteins and the carbonyl groups will form Schiff’s bases with the proteins that interfere with its structure.
CAS Number: 9002-93-1
Molecular formula: C34H62O11 for x=10
Molecular weight: 647 (for x=10)
Absorbance (254 nm): 0.16 (0.05% w/v)
Critical micelle concentration (CMC): ~0.2 × 10 to 3M (25 °C)
Aggregation number: 100 to 155
Cloud point: 65 °C
Average micellar weight: 80000
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.