BrainStorm STEM Earthquake Simulator Kit | Avantor
BrainStorm STEM Earthquake Simulator Kit
New Product
BrainStorm STEM Earthquake Simulator Kit
New Product
Certificates
BrainStorm STEM Earthquake Simulator Kit
BrainStorm STEM Earthquake Simulator Kit
2 Options Available Below
Product Details & Documents
Simulate seismic activity to explore structural integrity and earthquake science.
  • Designed by teachers, for teachers
  • Demonstrates structural design and stability
  • Perfect for home or the classroom
  • Available as a single kit or a class pack of ten with educator guide
  • No glue or mess
The BrainStorm earthquake simulator STEM kit helps students investigate structural integrity through realistic seismic testing. Learners build their own structures and test them on a vibrating tremor table to understand how buildings respond to different frequencies and intensities of movement. This activity enhances STEM concepts in physics and engineering. These kits are authenticated as educational kits by STEM.org.
Delivery information: Kit includes components for structure and tremor table, assembly instructions, and educator support materials.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • BrainStorm earthquake simulator STEM kit, class set
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $553.00
    $553.00
    Pack of 10
       $553.00
    La quantité minimale de commande est de 1
    Supplier Number: SK200-0005
    Spécifications:
    New Product
    New Product
  • BrainStorm earthquake simulator STEM kit, single
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $55.30
    $55.30
    Each
       $55.30
    La quantité minimale de commande est de 1
    Supplier Number: SK100-0005
    Spécifications:
    New Product