Periscope Kit
New Product
Build a periscope to learn about light reflection and line of sight.
Designed by teachers, for teachers
- Demonstrates angles of reflection and mirror alignment
- Perfect for home or the classroom
- Available as a single kit or a class pack of ten with educator guide
- No glue or mess
The BrainStorm Periscope STEM Kit invites students to construct and experiment with a functional periscope. Through hands-on learning, they explore how mirrors redirect light to see around corners, reinforcing principles of reflection and optics. These kits are authenticated as educational kits by STEM.org.
:
Kit includes pre-cut periscope parts, mirrors, adhesive, instruction manual.
Frequently Bought Together