Tornado Kit
Fournisseur: BRAINSTORM STUDIOS, LLC
New Product
Certificats
À propos de cet article
Simulate a swirling vortex to explore fluid dynamics and weather systems.
Designed by teachers, for teachers
- Demonstrates vortex formation and weather science
- Perfect for home or the classroom
- Available as a single kit or a class pack of ten with educator guide
- No glue or mess
The BrainStorm Tornado STEM Kit allows students to simulate a mini tornado using a specially designed chamber. Learners explore the principles of fluid dynamics, rotation, and atmospheric pressure while creating controlled vortexes. A thrilling and educational way to introduce weather science. These kits are authenticated as educational kits by STEM.org.
Delivery:
Kit includes vortex chamber, motor components, battery holder, instruction manual.
Frequently Bought Together