Electric Guitar Kit
New Product
Build a functional electric guitar to explore sound science and electronics.
Designed by teachers, for teachers
- Demonstrates principles of sound science and electronics
- Encourages hands-on STEM learning
- Perfect for home or the classroom
- Available as a single kit or a class pack of ten with educator guide
- No glue or mess
The BrainStorm Electric Guitar STEM Kit provides students with a unique opportunity to explore the science of sound and electronics. By building a functional electric guitar, learners can understand how vibrations and electrical signals produce music. This hands-on activity reinforces theoretical knowledge and enhances problem-solving skills. These kits are authenticated as educational kits by STEM.org.
:
Kit includes all necessary components to build the electric guitar and instruction manual. Class pack includes ten kits and an educator guide.
Frequently Bought Together