MicroPolx® 1900 Sealed Edge Cleanroom Wiper
Catalog # 77996-674
Supplier: Berkshire
New Product
Specifications
- Color:White
- Material:75% polyester/25% nylon
- Packaging:Double wrapped
- W×L:22.9×22.9 cm (9x9")
Specifications
About this item
MicroPolx® 1900 is a knitted polyester/nylon microfiber sealed edge cleanroom wiper featuring an ultra-fine microfiber yarn that increases surface area and provides superior wiping efficiency.
Removes oils and soils better than a traditional wiper
- Soft and non-abrasive fabric surface is excellent for sensitive or scratch prone surfaces
- Continuous filament yarns and tight knit construction provide excellent abrasion resistance and low fiber shedding
- Chemically compatible with IPA and other commonly used solvents
- Works well on both dry and wet applications
Ultra-fine, wedge-shaped polyester and nylon filaments, tight knit construction, and a non-abrasive smooth surface allows MicroPolx® 1900 to provide superior particle and bacterial removal without damaging critical surfaces. The high fiber surface area results in more efficient cleaning performance and minimizes the need for potentially harmful solvents.
Certifications:
ISO class 5 – 8. Cleanroom class 100 – 100000. EU Grade B/C/D.
Packaging:
Laundered and packaged in an ISO Class 5 (Class 100) cleanroom.