TubeWriter™ 400 UV Curable Inks
Supplier: TUBEWRITER, LLC
New Product
Certificates
About this item
TubeWriter's proprietary disposable UV curable ink is formulated to bond directly to any labware surface.
Bonds directly to any labware surface
Resistant to chemical solvents
Survives long-term storage at −80 °C and liquid nitrogen
Eliminates the need for sticky labels
Disposable cartridges with low replacement costs
To ensure the highest degree of solvent and temperatures protection possible, the ink has a shelf-life of 90 days.
The quarterly subscription delivers fresh ink cartridges every 90 days to ensure the highest degree of solvent and temperatures protection possible.