Kidder Pitching Machine STEM Build Kit
470358-012
New Product
- Description:Pitching machine STEM build kit
Construct a motorized pitching machine to explore basic electricity and mechanics.
- Encourages hands-on STEM learning Demonstrates principles of circuits and propulsion Includes four foam balls for safe indoor use Includes online teacher resource guide
The Pitching Machine STEM Build Kit offers students an engaging hands-on experience to explore the fundamentals of electricity and mechanical motion. By assembling a motorized pitching machine, learners gain insight into how electrical energy can be converted into mechanical energy to perform work. This activity not only reinforces theoretical knowledge but also enhances problem-solving skills, making it an excellent addition to STEM education programs.
Application: Engineering, Mechanics