Kidder Adjustable Stop Catapult | Avantor
Kidder Adjustable Stop Catapult
Adjustable stop catapult
Catalog # 470358-042
Supplier:   KIDDER MFG. CO. LTD
Fournisseur #:  83-K842461
Kidder Adjustable Stop Catapult
Catalog # 470358-042
Kidder Adjustable Stop Catapult
Adjustable stop catapult
Price
$102.00
Log in pour la disponibilité
Kit of 1
La quantité minimale de commande est de 1
Product Details & Documents
Construct a working catapult with adjustable stop for trajectory experiments.
  • Includes adjustable stop for range control
  • Explores projectile motion and energy transfer
  • Reinforces principles of physics and design
  • Compact and reusable for multiple trials
The Adjustable Stop Catapult kit enables students to investigate projectile motion, force, and energy through hands-on experimentation. With a modifiable stop point, learners can adjust the launch angle and observe how it affects trajectory. This activity supports a wide range of physics lessons and promotes inquiry-based learning through design and testing.
Application: Engineering, Mechanics
Spécifications
  • Désignation:
    Adjustable stop catapult