Kidder Adjustable Stop Catapult
Adjustable stop catapult
Catalog # 470358-042
Supplier: KIDDER MFG. CO. LTD
Fournisseur #: 83-K842461
Jump to:
Product Details & Documents
Construct a working catapult with adjustable stop for trajectory experiments.
- Includes adjustable stop for range control
- Explores projectile motion and energy transfer
- Reinforces principles of physics and design
- Compact and reusable for multiple trials
The Adjustable Stop Catapult kit enables students to investigate projectile motion, force, and energy through hands-on experimentation. With a modifiable stop point, learners can adjust the launch angle and observe how it affects trajectory. This activity supports a wide range of physics lessons and promotes inquiry-based learning through design and testing.
Application: Engineering, Mechanics
Spécifications
- Désignation:Adjustable stop catapult