ContactUsLinkComponent
Kidder Rope Climbing Robot STEM Build Kit
  470358-018
New Product
undefined
Kidder Rope Climbing Robot STEM Build Kit
  470358-018
 :  80-54-W051
New Product
  • Description:
    Rope climbing robot STEM build kit
Assemble a robot that climbs rope using pulley-based motion.
  • Demonstrates mechanical advantage and pulley systems
  • Uses motorized climbing action
  • Hands-on STEM building activity
  • Ideal for engineering and motion lessons
The Rope Climbing Robot STEM Build kit introduces students to simple machines and mechanical advantage through an engaging project. As students assemble and observe the motorized robot climbing a rope, they gain insight into how pulleys and gears convert energy into motion. This hands-on kit is perfect for exploring motion, energy transfer, and basic robotics concepts in a tangible way.
Application: Engineering