BCR® Flat Mop Covers, Polyester
About this item
Polyester flat mop covers are available in non sterile or sterile (irradiated) formats.
Abrasion-resistant polyester knit cleanroom fabric laminated to absorbent polyurethane foam
- Laser cut to minimize fiber and particle release
- 10 mm thickness provides cushioned surface for optimum conformance to uneven floors
- High absorbency capacity
- Compatible with a wide range of chemicals, disinfectants, and cleaners
- Autoclavable and compatible with gamma and ETO sterilization
Choose from a variety of mop covers compatible with the BCR® all-in-one stainless steel mop cart system, each designed for efficient cleaning of floors, walls, and other large surfaces critical to the overall cleanliness of the production environment.
Certifications: ISO Class 3 – 8. Cleanroom Class 1 – 100000. EU Grade B/C/D. USP 797.
2 Options Available Below
Product Details & Documents
Polyester flat mop covers are available in non sterile or sterile (irradiated) formats.
Abrasion-resistant polyester knit cleanroom fabric laminated to absorbent polyurethane foam
- Laser cut to minimize fiber and particle release
- 10 mm thickness provides cushioned surface for optimum conformance to uneven floors
- High absorbency capacity
- Compatible with a wide range of chemicals, disinfectants, and cleaners
- Autoclavable and compatible with gamma and ETO sterilization
Choose from a variety of mop covers compatible with the BCR® all-in-one stainless steel mop cart system, each designed for efficient cleaning of floors, walls, and other large surfaces critical to the overall cleanliness of the production environment.
Certifications: ISO Class 3 – 8. Cleanroom Class 1 – 100000. EU Grade B/C/D. USP 797.
Product Family Options
Product Information
- DimensionsMatièreSterilityColorPackedAvailabilityPrice
- 40×12 cm (16×5")Polyester knit/polyurethane foamNon sterileWhiteStackedLog in for availabilityCase of 100$569.00Supplier Number: FMCPET20Specifications:New ProductNew Product
- 40×12 cm (16×5")Polyester knit/polyurethane foamSterileWhiteStackedLog in for availabilityCase of 100$722.00Supplier Number: FMCPET20IRSpecifications:New ProductNew Product