Counstar Spica M1 Real-Time Live Cell Imaging and Analysis System
Catalogue # 77973-670
Fournisseur: ALIT LIFETECH INC
New Product
Spécifications
- Description:Real-time live cell imaging and analysis platform for 2D/3D cultures
- W×D×H:12×12×13"
- Camera:7.1 MP, CMOS
- Magnification:7.5×, 10×, 15×
- Wavelength:Ex: 375/28 nm, Em: 430/30 nm; Ex: 470/20 nm, Em: 515/40 nm; Ex: 560/25 nm, Em: 630/70 nm (fluorescent channels)
- Weight:20 lbs.
Spécifications
À propos de cet article
The Countstar Spica provides real-time live cell imaging and analysis to support a wide variety of applications in life science research, drug development, and cell-based therapies.
Incubator-friendly for optimal cell health
- Monitor experiments from anywhere, anytime for maximu flexibility and efficiency
- Ready-to-go assays with pre-set parameters for consistent and reliable data for all users
- Seamless imaging to analysis in one software
- Unparalleled flexibility for diverse live cell assays
The integration of advanced scanning technology and AI-powered image processing algorithms enables fast and accurate analyses of both 2D cell cultures and 3D organoids. The comprehensive suite of workflow solutions simplifies operation while providing the flexibility to customize and expand the scope of applications to meet new challenges.