Soil Test Kit | Avantor
Soil Test Kit
Physical soil test kit
Catalog # 470358-648
New Product
Supplier:   Luster Leaf
Fournisseur #:  1662
Soil Test Kit
Catalog # 470358-648
Supplier:  Luster Leaf
New Product
Soil Test Kit
Soil Test Kit
Physical soil test kit
Price
$47.32
Log in pour la disponibilité
Pack of 40 Case of 240
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Quickly and easily tests soil samples measuring levels of pH, nitrogen, phosphorus, and potash.
  • Easy to use
  • Compare test results against colorimetric and turbidity reading charts
Your students become environmental scientists with this hands-on real world product. Test the soil at your school or in your flower pots. The tests are performed in four color coded test tubes, one each for pH, nitrogen, phosphorus, and potash. A dropper is provided to facilitate transfererring the test solution into the test tubes. The kit also includes a pamphlet with pH preferences for hundreds of plants, including vegetables, herbs, flowers, trees, shrubs, fruits, grasses, and house plants.
Delivery information: Includes special cleaning pad to be used in between each test.
Spécifications
  • Désignation:
    Physical soil test kit