Rubbermaid® Commercial Low Profile Scrub-Strip Carpet Bonnet
77929-394
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Color:White/green
- Description:Low profile scrub-strip carpet bonnet
- Size:21" Ø
Low profile carpet bonnets with green scrubber strip makes floor cleaning easy.
- Includes green scrubber strips for stubborn blemishes Standard 21" Ø Use with shampoo or solvent based chemicals
This 21" carpet bonnet is intended for use with 175-300 rpm floor machines and features scrubber strips to smoothly remove stubborn blemishes with minimal effort. For use with shampoo or solvent based chemicals.