Five Star® Composition Book, Medium/College Rule
Catalog # 77929-436
Supplier: Janitorial Supplies
New Product
Specifications
- Description:Composition book, medium/college rule
Specifications
About this item
A Five Star® Composition Book is perfect for permanent notetaking. Use it in class and at home for labs, take-home assignments, and more.
Composition book features wireless sewn binding to keep pages permanently bound
- Durable, plastic cover lasts all year and lies flat when opened
- Class schedule printed inside
- Randomly assorted cover color
- (80) 9.75×7.5" sheets
These composition books are made to be durable, eliminating the worry about binding coming apart or missing pages that have fallen out. Pages are sewn in so they stay in place securely, all year long. The composition book front and back covers are plastic to resist everyday wear and tear. Use with Five Star® Study App to scan, study, and organize notes. Create instant flashcards and sync notes to Google Drive to access them anywhere from any device.