ContactUsLinkComponent
Boardwalk® Stain-Resistant Floor Sealer, Unscented
Boardwalk® Stain-Resistant Floor Sealer, Unscented
Catalog # 77929-418
New Product
undefined
Boardwalk® Stain-Resistant Floor Sealer, Unscented
Catalog # 77929-418
Supplier Number:  BWK4404SL
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Stain-resistant floor sealer
  • Size:
    1 gal.
  • Packaging:
    Bottle
Specifications
About this item
Floor sealer works well as a base coat before applying finish.
  • Protects against asphalt, petroleum, and common food and drink stains
  • Increases gloss of subsequently applied top coats on porous industrial floors
  • Unscented
Floor sealer protects floors against common food and drink stains, petroleum, and asphalt. Increases gloss of subsequently applied top coats on porous industrial floors. Stain-resistant floor sealer creates a level, clear gloss surface for high-traffic vinyl and other hard surface floors. Provides excellent value; one gallon covers approximately 1500 square feet.