Diversey™ Wiwax ProX Maintenance Emulsion
Catalog # 77929-438
Supplier: Janitorial Supplies
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Wiwax ProX maintenance emulsion
- Packaging:Bottle
Specifications
About this item
An integral part of a comprehensive floor care program for cleaning, stripping, and sealing rubber floors.
Provides the floor with a shiny appearance
- Provides protection when used on a regular basis
Wiwax ProX Maintenance emulsion is a unique blend of waxes, polymers, and detergents to effectively rejuvenate and maintain rubber as well as linoleum, asphalt, marble, stone, concrete, and slate flooring.