ContactUsLinkComponent
Energizer® Industrial Lithium CR2 Photo Batteries, 3 V
Catalogue # 77929-444
Fournisseur:  Janitorial Supplies
New Product
undefined
Energizer® Industrial Lithium CR2 Photo Batteries, 3 V
Catalogue # 77929-444
Fournisseur:  Janitorial Supplies
Supplier Number:  EVEELN1CR28
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Energizer® industrial lithium CR2 photo batteries, 3V
Spécifications
À propos de cet article
Rely on energizer industrial lithium CR2 batteries to power specialty professional devices.
  • Keep all specialty devices powered up for work
  • Safeguard professional equipment with lithium batteries that are 100% leakproof in standard usage
  • Get remarkable performance, even in extreme work site temperatures
Trust your precision electronics to lithium CR2 batteries that are 100% leakproof in normal usage. They operate in work site temperatures ranging from −40 to +140 °F. They hold their power with up to ten years of shelf life, virtually eliminating device downtime. When work depends on laser pointers, motion sensors, automatic paper towel dispensers, and more, rely on the power and protection of energizer industrial lithium CR2 batteries.