Dynamics Track System | Avantor
Dynamics Track System
Supplier:   GSC International
Dynamics Track System
Supplier:  GSC International
Certificates
Dynamics Track System
Dynamics Track System
2 Options Available Below
Product Details & Documents
This cost-effective dynamics system is a versatile tool for studying linear motion and collisions.
  • Available with and without photogates
  • Comes with a large assortment of accessories to facilitate many different types of experiments
A dynamic cart and track system is an advanced physics lab tool used to study motion, forces, momentum, and energy with high-precision timing measurements. Measure velocity, acceleration, and collision events with high accuracy. Newton’s Laws experiments - Demonstrate inertia, force, and action-reaction principles. Momentum and Energy Conservation - Study elastic and inelastic collisions with real-time data collection. Kinematic studies - Analyze velocity, acceleration, and free-fall motion with detailed measurements. Friction and Inclined Plane investigations - Examine how forces and motion change with different surfaces or slopes.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Dynamics track system
    120 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $711.00
    $711.00
    Each
       $711.00
    La quantité minimale de commande est de 1
    Supplier Number: DCTS-1
  • Dynamics track system with photogates
    120 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $971.70
    $971.70
    Each
       $971.70
    La quantité minimale de commande est de 1
    Supplier Number: DCTS-2