ColorView™ Histology Slides
Fournisseur: StatLab
New Product
Certificats
À propos de cet article
Slides for routine H&E or special stain applications.
ColorView™ non-adhesion slides are an excellent, cost-effective choice for routine H&E and special stain procedures.
Packaging:
Slides are packaged 10 gross per case.
Frequently Bought Together