VWR® Autoclavable Biohazard Bags, Polypropylene, Yellow
Fournisseur: VWR International
New Product
Certificats
À propos de cet article
Sturdy, temperature-resistant bags are ideal for disposal of non-hazardous waste that must first be autoclaved. Bags can also be used for autoclaving other products.
Autoclavable up to 135 °C (275 °F) maximum
Printed bags feature the universal biohazard symbol
Temperature indicator text ‘Contents Have Been Autoclaved’ turns dark to indicate successful autoclaving
Highly resistant to tears and punctures
Bags are supplied complete with 20 cm twist tie wire closures that allow expansion of the bag and ventilation when cooling. Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709.
Certifications:
ISO 9001:2015, Sedex.
Frequently Bought Together