ContactUsLinkComponent
Genecon Static Electricity Generator
Catalog # 470357-008
Supplier:  GSC International
New Product
Genecon Static Electricity Generator
Catalog # 470357-008
Supplier:  GSC International
Supplier Number:  N-00029
New Product
Specifications
  • Description:
    Genecon static electricity generator
Specifications
About this item
Handheld generator producing static electricity through friction, ideal for classroom experiments.​
  • Transparent design allows users to observe the internal mechanism, demonstrating electricity generation without motors or batteries
  • Operates safely in various environments, including high humidity and heat
  • Enables a variety of static electricity experiments, illustrating concepts like charge polarity and electrostatic induction
  • Designed with durable materials; replacement gear sets are available, reducing long-term costs
The Genecon static electricity generator is a handheld device designed to generate static electricity through the friction of two different materials, specifically polyvinyl chloride (PVC) resin and felt. Its transparent polycarbonate housing allows users to observe the internal workings, providing a clear demonstration of electricity generation without the use of motors or batteries. This feature enhances understanding of the triboelectric series and electrostatic induction principles.
The device is capable of producing more than 10000 V at the output end of the dedicated cables, making it suitable for a wide range of static electricity experiments in educational settings. Its design ensures safe operation even in high humidity and hot conditions, addressing common challenges associated with traditional static electricity generators.
Additionally, the Genecon static electricity generator is constructed with durable materials, including polyacetal resin gears, and offers cost-effective maintenance options through the availability of replacement gear sets.