Miniature Daniell Cell, Basic Set
Catalog # 470357-028
Supplier: GSC International
New Product
Specifications
- Description:Mini Daniell cell, basic set
Specifications
About this item
A compact, hands-on kit designed for microscale experiments demonstrating electrochemical principles with minimal chemical usage.
Utilizes a semipermeable membrane instead of a traditional porous cup or salt bridge
- Minimizes the use of chemicals and reduces waste fluid. This makes it an environmentally considerate choice for classroom and laboratory use
- Designed to provide practical understanding of ion exchange, electron flow, and redox reactions, bridging theoretical knowledge with real-world applications
Delivery:
Kit includes carrying case; (4) 10 ml drop bottles; (25) filter papers (60×90 mm); (40) cellophane pieces (50×50 mm); (6) electrode screws (37 mm); (6) cork plugs; (4) plastic cylinders; (6) rubber rings; (20) metal plates (10×10×1 mm), five each of Cu, Zn, Fe, Al.