ContactUsLinkComponent
Innovating Science® Synthesis of a Coordination Complex Lab Kit
Catalogue # 470357-890
Fournisseur:  Ward's Science
New Product
undefined
Innovating Science® Synthesis of a Coordination Complex Lab Kit
Catalogue # 470357-890
Fournisseur:  Ward's Science
Supplier Number:  IS2535
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Synthesis of a coordination complex lab kit
  • No. of Lab Groups:
    15
Spécifications
À propos de cet article
Produce a bright-green iron compound in this multi-step synthesis.
  • Kit contains enough materials for 15 groups Teacher's manual and student study guide copymaster included
Coordination complexes are brightly colored compounds consisting of a central metal atom and ligand molecules, atoms, or ions. Synthesize and purify a lime-green coordination complex called potassium ferrioxalate trihydrate in this multi-step synthesis and optionally analzye the complex via UV-Visible spectrophotometry.
Ordering information: Required but not included: 150 ml Glass beakers, funnels, hot plate stirrers, magnetic stir bars, thermometers, distilled or deionized water, laboratory scale, 25 or 50 ml graduated cylinders, tongs or heat-resistant gloves, forceps, spatulas, ice bath, 500 ml glass beakers, cuvettes [optional], UV-Visible spectrophotometer [optional].
Delivery: Kit includes: Oxalic acid 0.5 M solution, 800 ml; Potassium oxalate 1.0 M solution, 225 ml; Hydrogen peroxide 3% solution, 225 ml; Ethanol, 95%, 150 ml; Ethanol, 50%, 225 ml; Sulfuric acid, 0.1 M, 25 ml; Ferrous ammonium sulfate hexahydrate, 50 g; 30× pipettes; 15× filter papers.