Innovating Science® Introduction to Complexometric Titration Lab Kit
Catalog # 470357-876
Supplier: Ward's Science
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Introduction to complexometric titration lab kit
- No. of Lab Groups:15
Specifications
About this item
Perform a common analytical technique, complexometric titration, to quantify calcium in samples.
- Kit contains enough materials for 15 groups Teacher's manual and student study guide copymaster included
Complex formation is frequently used in both qualitative and quantitative inorganic analysis. Introduce topics such as metal-ligand interactions in this activity using complexometric titration to determine the calcium content of both known and unknown samples.
Ordering information: Required but not included: 125 ml Erlenmeyer flasks, 50 ml burettes, ring stands with burette clamps, 10 or 25 ml graduated cylinders, funnels, distilled or deionized water, white paper [optional], magnetic stir plates and stir bars [optional].
Delivery: Kit includes: 4× EDTA 0.1 M solution, 500 ml; 3× Eriochrome black T (EBT) indicator solution, 60 ml; Calcium chloride 0.1 M solution, 500 ml; unknown Calcium solution #1, 500 ml; unknown Calcium solution #2, 500 ml; 3× pH 10 buffer solution 500 ml.